Nylon Tube Brushes and Pipe Cleaner, Restek
# de catalogue RK20108
Fournisseur: Restek
Spécifications
- Bristle Material:Nylon
- Description:Nylon Tube Brushes and Pipe Cleaner
- Cat. no.:RK20108
- Supplier No.:20108
Spécifications
A propos de cet article
Use nylon tube brushes and pipe cleaners to remove small septum fragments and residue from dirty glass inlet liners.
- Brushes and pipe cleaners are available in different diameters
- Useful for fine cleaning operations
Brushes are 1/8, 3/16 and 1/4" in diameter; pipe cleaner is one foot (30 cm) long.
Customers Who Bought This Also Bought