ContactUsLinkComponent
Capillary Column Nylon Tube Brushes and Pipe Cleaner
Nylon Tube Brushes and Pipe Cleaner, Restek
# de catalogue RK20108
Fournisseur:  Restek
Capillary Column Nylon Tube Brushes and Pipe Cleaner
Nylon Tube Brushes and Pipe Cleaner, Restek
# de catalogue RK20108
Fournisseur:  Restek
Spécifications
  • Bristle Material:
    Nylon
  • Description:
    Nylon Tube Brushes and Pipe Cleaner
  • Cat. no.:
    RK20108
  • Supplier No.:
    20108
Spécifications
A propos de cet article
Use nylon tube brushes and pipe cleaners to remove small septum fragments and residue from dirty glass inlet liners.
  • Brushes and pipe cleaners are available in different diameters
  • Useful for fine cleaning operations
Brushes are 1/8, 3/16 and 1/4" in diameter; pipe cleaner is one foot (30 cm) long.
Customers Who Bought This Also Bought