Hydrochloric acid 36.5 - 38.0%, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # JT9535-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7647-01-0
MOC: Non-Regulated, With InformationCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Glass bottle, clear
- Size:500 ml
- Supplier No.:9535-01
Specifications
Specification Test Results
- Appearance:Passes Test
- Assay (as HCl) (by acid-base titrn):36.5 - 38.0 %
- Color (APHA):≤ 10
- Extractable Organic Substances:≤ 5 ppm
- Free Chlorine (as Cl₂):≤ 1 ppm
- Residue after Ignition:≤ 3 ppm
- Specific Gravity at 60°/60°F:1.185 - 1.192
- Bromide (Br):≤ 0.005 %
- Phosphate (PO₄):≤ 1 ppm
- Sulfate (SO₄):≤ 0.5 ppm
- Sulfite (SO₃):≤ 0.8 ppm
- Ammonium (NH₄):≤ 3 ppm
- Trace Impurities - Aluminum (Al):≤ 100.0 ppb
- Arsenic and Antimony (as As):≤ 5.0 ppb
- Trace Impurities - Boron (B):≤ 50 ppb
- Trace Impurities - Calcium (Ca):≤ 200.0 ppb
- Trace Impurities - Chromium (Cr):≤ 100.0 ppb
- Trace Impurities - Copper (Cu):≤ 100.0 ppb
- Trace Impurities - Gold (Au):≤ 100.0 ppb
- Heavy Metals (as Pb):≤ 100 ppb
- Trace Impurities - Iron (Fe):≤ 100.0 ppb
- Trace Impurities - Lead (Pb):≤ 50 ppb
- Trace Impurities - Magnesium (Mg):≤ 300.0 ppb
- Trace Impurities - Manganese (Mn):≤ 300.0 ppb
- Trace Impurities - Mercury (Hg):≤ 5.0 ppb
- Trace Impurities - Nickel (Ni):≤ 100.0 ppb
- Trace Impurities - Potassium (K):≤ 300.0 ppb
- Trace Impurities - Sodium (Na):≤ 300 ppb
- Trace Impurities - Tin (Sn):≤ 300.0 ppb
- Trace Impurities - Titanium (Ti):≤ 300.0 ppb
- Trace Impurities - Zinc (Zn):≤ 100.0 ppb
Specification Test Results
About this item
Synonyms: Hydrochloric acid solution
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–35 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
:Management of Change (MOC) category = R
EINECS:231-595-7
:GHS05
:GHS07
Frequently Bought Together