Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
# de catalogue BDH1172-4LP
Fournisseur: VWR International
Numéro CAS: 56-81-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle
- Size:4 L
- Supplier No.:PL380ZA-4P
Spécifications
Résultats des tests de spécification
- Appearance:Clear
- Identification:Passes test
- Water (K.F.):Less than 0.3%
- Residue after Ignition:Less than 0.01%
- Assay (calc from Specific Gravity, use % Glycerine test):Greater than 99.7%
- Color (APHA):Less than 20
- Specific Gravity (25ºC/25ºC):1.2613 or greater
- Chlorides:Less than 10 ppm
- Sulfates:Less than 20 ppm
- Heavy Metals (as Pb):Less than 5 ppm
- Chlorinated Compounds (as Cl):Less than 30 ppm
- Fatty Acids and Esters:Passes test
- Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®:
Résultats des tests de spécification
A propos de cet article
Synonymes: 1,2,3-Propanetriol,Propane-1,2,3-triol
Point d'ébullition:290 °C (1013 hPa)
Point de Fusion:18.6 °C
Densité:1.26 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,7
Point d'éclair:177 °C
:Boiling Pt:
:Boiling Pt: 290 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt: 177 °C ()
:M.W. 92.09 g/mol
Index Merck:13,04497
Température d'entreposage:Ambient
EINECS:200-289-5
:177
Customers Who Bought This Also Bought