2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Catalog # BDH1133-19L
Supplier: VWR International
CAS Number: 67-63-0
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Steel can
- Size:19 L
- Cat. no.:BDH1133-19L
- Supplier No.:PL323ZA-19
Specifications
Specification Test Results
- Appearance:Clear
- Water (K.F.):Less than 0.20%
- Residue:Less than 0.001% (0.0010 g/128 mL)
- GC Purity:Greater than 99.5%
- Solubility in water:Passes test (Solution should be as clear as an equal volume of water.)
- Color (APHA):Less than 10
- Titrable acid or base:Less than 0.1 meq/g (No more than 0.10 mL of either 0.01N sodium hydroxide or 0.01N hydochloric acid)
- Carbonyl compounds:Less than 0.002% each (as acetone or propionaldehyde)
- VWR Chemicals BDH®, 2-Propanol ≥99.5% ACS:
Specification Test Results
About this item
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
Flash Pt:12 °C
:Boiling Pt:
:Boiling Pt: 82 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –89 °C
:Flash Pt:
:Flash Pt: 12 °C
:M.W. 60.1 g/mol
Merck Index:13,05228
:2-Propanol
:GHS02
:GHS07
:3
:II
UN:1219
:82
:12
:-89
Storage Temperature:Ambient
EINECS:200-661-7
REACH:01-2119457558-25
Customers Who Bought This Also Bought