Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®
# de catalogue BDH1109-4LG
Fournisseur: VWR International
Product available on GSA Advantage®
Numéro CAS: 67-66-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:BDH1109-4LG
- Supplier No.:PL049ZA-4
Spécifications
Résultats des tests de spécification
- Appearance:Clear, colorless liquid
- Water (K.F.):Less than 0.020%
- Residue:Less than 0.001% (0.0010g/67 mL)
- GC Purity:Greater than 99.8%
- Amylene (Capillary GC-FID):Record value
- Color (APHA):Less than 10
- Acetone and aldehyde:Passes test (less than 0.005% as acetone. No turbidity should develop.)
- Acid and chloride:Passes test (Blue litmus paper should not change color. No turbidity should be produced.)
- Free chlorine (Cl):Passes test (Lower layer should not show a violet tint.)
- Lead (Pb):Less than 0.05 ppm (Purplish hue of sample should not exceed that of the standard.)
- VWR Chemicals BDH®, Chloroform ≥99.8% stabilized ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Methane, trichloro-,Trichloroform
Point d'ébullition:61.7 °C (1013 hPa)
Point de Fusion:–63 °C
Densité:1.4832 g/cm³ (20 °C)
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Stabilised with amylene
:Boiling Pt:
:Boiling Pt: 61.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –63 °C
:Flash Pt:
:Flash Pt:
:M.W. 119.38 g/mol
Index Merck:13,02160
EINECS:200-663-8
:-63
ONU:1888
:05-2114614906-45
:6.1
:III
:GHS06
:GHS08
Température d'entreposage:Ambient
:Amylen
Customers Who Bought This Also Bought