ContactUsLinkComponent
40% Solution Containing 38.96% (W/V) Acrylamide And 1.04% (W/V) Bis-Acrylamide
VWR® Acrylamide and Bis-Acrylamide
# de catalogue 97064-542
Fournisseur:  VWR
40% Solution Containing 38.96% (W/V) Acrylamide And 1.04% (W/V) Bis-Acrylamide
VWR® Acrylamide and Bis-Acrylamide
# de catalogue 97064-542
Fournisseur:  VWR
Spécifications
  • Description:
    Acryl/Bis™ 37.5:1
  • Size:
    500 mL
  • Supplier No.:
    0254-500ML
Spécifications
A propos de cet article
An extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, premixed stock solutions and ready-to-use solutions for customized polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids. Ultra-pure (>99.9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimize the handling of neurotoxic acrylamide.
An extensive line of acrylamide and bis-acrylamide pre-weighed powder blends, premixed stock solutions and ready-to-use solutions for customized polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids. Ultra-pure (>99.9%) acrylamide and bis-acrylamide powders provide the flexibility to prepare solutions having concentrations and ratios for all electrophoresis applications. Liquid blends minimize the handling of neurotoxic acrylamide.
Customers Who Bought This Also Bought