ContactUsLinkComponent
AGAROSE RA 500GM
VWR® Agarose RA™ Agarose RA
Catalog # 97064-258
Supplier:  VWR
AGAROSE RA 500GM
VWR® Agarose RA™ Agarose RA
Catalog # 97064-258
Supplier:  VWR
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    500 g
  • Cat. no.:
    97064-258
  • Supplier No.:
    N605-500G
Specifications
About this item
A quality general application agarose that provides good resolution and is cost-effective for high volume users.
  • Ideal Agarose for High Throughput
  • High Resolution
  • Easy Preparation
  • Excellent Sensitivity
A quality general application agarose that provides good resolution and is cost-effective for high volume users. It was developed specifically for quick checks of PCR products, plasmids preps, screening and cloning. Features high resolution, easy preparation and set up, a low EEO and excellent sensitivity with low background when stained.
Customers Who Bought This Also Bought