Specifications
- Size:500 g
- Cat. no.:97064-258
- Supplier No.:N605-500G
Specifications
About this item
A quality general application agarose that provides good resolution and is cost-effective for high volume users.
- Ideal Agarose for High Throughput
- High Resolution
- Easy Preparation
- Excellent Sensitivity
A quality general application agarose that provides good resolution and is cost-effective for high volume users. It was developed specifically for quick checks of PCR products, plasmids preps, screening and cloning. Features high resolution, easy preparation and set up, a low EEO and excellent sensitivity with low background when stained.