VWR® Phosphate Buffered Saline (PBS), 10X, Sterile, Ultra Pure Grade
# de catalogue 97063-660
Fournisseur: VWR
Product available on GSA Advantage®
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Size:500 mL
- Supplier No.:K813-500ML
Spécifications
A propos de cet article
1X Solution Composition: 137 mM NaCl, 2.7 mM KCl, 9.5 mM Phosphate buffer.
- Dilute 1:10 in sterile water for making 1X solution
- 10X solution is supplied 0.2 um filter sterilized
- pH at 25°C (1X PBS): 6.6 – 7.2
- DNase, RNase, Protease none detected
- Bioburden Pass (USP)
- Suitable for cell culture
- Does not contain magnesium or calcium
Customers Who Bought This Also Bought