Order Entry
ContactUsLinkComponent
BUFFER TBE 5X LIQ CONC ULT-PR GRD 1L
VWR® TRIS-Borate-EDTA Buffer, 5X Concentrate, Ultra Pure Grade
# de catalogue 97063-432
Fournisseur:  VWR
Numéro CAS:  
BUFFER TBE 5X LIQ CONC ULT-PR GRD 1L
VWR® TRIS-Borate-EDTA Buffer, 5X Concentrate, Ultra Pure Grade
# de catalogue 97063-432
Fournisseur:  VWR
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Size:
    1 L
  • Cat. no.:
    97063-432
  • Supplier No.:
    J885-1L

Spécifications

A propos de cet article

TBE is optimized for DNA electrophoresis techniques using both acrylamide and agarose

  • High Buffering Capacity
  • High Ionic Strength
  • Permits Extended Electrophoresis Runs with no Buffer Recirculation
  • Convenient Packaging Options Minimize Weighing and Mixing

After dilution the composition of (1X) Solution contains 0.089 M Tris Base, 0.089 M Borate and 0.002 M EDTA, Disodium Salt, Dihydrate; final pH 8.3.

Customers Who Bought This Also Bought