Bovine Serum Albumin (BSA)
# de catalogue 97062-904
Fournisseur: VWR
Numéro CAS:
Spécifications
- Packaging:HDPE Bottle
- Source:Serum
- Species:Bovine
- CAS No.:9048-46-8
90604-29-8 - Size:100 g
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Colour:light yellow
- Grade:for radioimmunoassay (RIA)
- Supplier No.:E588-100G
Spécifications
A propos de cet article
Customers Who Bought This Also Bought