Reagent Reservoirs, VistaLab
# de catalogue 14007-066
Fournisseur: VistaLab
Spécifications
- Capacity:100 ml
- Sterility:Sterile
- Description:One-Compartment Tray with Lid
- Cat. no.:14007-066
- Packaging:Individually Wrapped
- Supplier No.:3054-3001
Spécifications
A propos de cet article
VistaLab reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery.
- Trough-within-a-trough design for greatest sample recovery
- Pour-off spouts at every corner
- Wider base for stability
- Free of animal products
- Sterile reservoirs are RNase/DNase-free and non-pyrogenic
- Bulk or individually wrapped
- 12-channel polypropylene reservoir is autoclavable
Sterile and non-sterile reservoirs and reservoir coolers are available for purchase, with bulk or individually wrapped formats for compatibility with any distribution method.
Customers Who Bought This Also Bought