ContactUsLinkComponent
RESERVOIR W/LID RGT 100ML EA50
Reagent Reservoirs, VistaLab
# de catalogue 14007-066
Fournisseur:  VistaLab
RESERVOIR W/LID RGT 100ML EA50
Reagent Reservoirs, VistaLab
# de catalogue 14007-066
Fournisseur:  VistaLab
Spécifications
  • Capacity:
    100 ml
  • Sterility:
    Sterile
  • Description:
    One-Compartment Tray with Lid
  • Cat. no.:
    14007-066
  • Packaging:
    Individually Wrapped
  • Supplier No.:
    3054-3001
Spécifications
A propos de cet article
VistaLab reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery.
  • Trough-within-a-trough design for greatest sample recovery
  • Pour-off spouts at every corner
  • Wider base for stability
  • Free of animal products
  • Sterile reservoirs are RNase/DNase-free and non-pyrogenic
  • Bulk or individually wrapped
  • 12-channel polypropylene reservoir is autoclavable
Sterile and non-sterile reservoirs and reservoir coolers are available for purchase, with bulk or individually wrapped formats for compatibility with any distribution method.
Customers Who Bought This Also Bought