Low-Pressure Slip-On Inlet Filter for Mobile Phase Reservoir, Restek
# de catalogue RK25008
Fournisseur: Restek
Spécifications
- Description:Low-Pressure Inlet Filter, 1/8" 10 µm Slip-On
- Cat. no.:RK25008
- Supplier No.:25008
Spécifications
A propos de cet article
A 316 stainless steel tip with a Tefzel® collar seals to a corrosion-resistant 316 stainless steel filter element.
The slip-on filter easily attaches to the pump inlet line, without the use of wrenches. The universal ¹/₈" Ext.Ø tip accommodates standard PTFE tubing inner diameters. The cylindrical filter is standard 10 µm porosity. Fits Altex, ISCO, LDC, Varian, Waters, PerkinElmer, and other pumps.
Customers Who Bought This Also Bought