Caps and Septa for 2.0 ml Big Mouth Step Vials, Preassembled, Restek
# de catalogue RK24677
Fournisseur: Restek
Spécifications
- Description:Caps and Septa, Preassembled
- Cat. no.:RK24677
- Supplier No.:24677
- Orifice:10 mm/425
Spécifications
A propos de cet article
Caps and septa for 2.0mL big mouth vials are designed to reduce needle-breaking and fit screw-thread vials.
- Screw-thread vial compatibility
- Economical
- Red PTFE/Silicone
Extra-wide 10 mm openings decrease the number of broken needles with a larger target area.
Ordering information: Choose preslit caps (available for some vials) to reduce the risk of needle bending, release vacuum from high-volume injections, and improve injection reproducibility when greater than 20% of vial volume is withdrawn.
Customers Who Bought This Also Bought