MINICYL Regulator, Restek
Catalog # RK20611
Supplier: Restek
Specifications
- Description:MINICYL Regulator, 1/4" Brass Fittings
- Cat. no.:RK20611
- Supplier No.:20611
Specifications
About this item
This compact general purpose regulator has many laboratory applications including air-drying glassware, sparging or evaporating solutions, and controlling pneumatic valves.
- Lightweight aluminum construction
- Maximum pressure of 300 psi
Customers Who Bought This Also Bought