ContactUsLinkComponent
MINICYL Regulator, 1/4" fittings
MINICYL Regulator, Restek
Catalog # RK20611
Supplier:  Restek
MINICYL Regulator, 1/4" fittings
MINICYL Regulator, Restek
Catalog # RK20611
Supplier:  Restek
Specifications
  • Description:
    MINICYL Regulator, 1/4" Brass Fittings
  • Cat. no.:
    RK20611
  • Supplier No.:
    20611
Specifications
About this item
This compact general purpose regulator has many laboratory applications including air-drying glassware, sparging or evaporating solutions, and controlling pneumatic valves.
  • Lightweight aluminum construction
  • Maximum pressure of 300 psi
Customers Who Bought This Also Bought