Septum Puller, Restek
# de catalogue RK20117
Fournisseur: Restek
Spécifications
- Description:Septum Puller
- Cat. no.:RK20117
- Supplier No.:20117
Spécifications
A propos de cet article
Septum pullers lend an enhanced level of comfort in easily removing septa and O-rings from GC systems.
- Effective at removing septa
- Easily clear debris with fine pointed end
Use hooked end for removing septa and O-rings. The pointed end works well for removing stuck ferrules or debris.
Customers Who Bought This Also Bought