Glycerine ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
# de catalogue MK509216
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 56-81-5
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Poly bottle
- Size:4 L
- Supplier No.:5092-16
Spécifications
Résultats des tests de spécification
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (C₃H₅(OH)₃) (by GC, corrected for water):≥99.5 %
- ACS - Acrolein and Glucose:Passes Test
- ACS - Chlorinated Compounds (as Cl):≤0.003 %
- ACS - Color (APHA):≤10
- ACS - Fatty Acid Esters (as Butyric Acid):≤0.05 %
- ACS - Heavy Metals (as Pb):≤2 ppm
- ACS - Neutrality:Passes Test
- ACS - Residue after Ignition:≤0.005 %
- Solubility (5 in 50):Passes Test
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Sulfate (SO₄):≤0.001 %
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.5 %
- Appearance (clear, colorless, syrupy liquid):
- Macron Fine Chemicals™, Glycerine ≥99.5% (by GC, corrected for water content), AR® ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: 1,2,3-Propanetriol,Propane-1,2,3-triol
Point d'ébullition:290 °C (1013 hPa)
Point de Fusion:18.6 °C
Densité:1.26 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = N
Point d'éclair:177 °C
:Boiling Pt:
:Boiling Pt: 290 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt: 177 °C ()
:M.W. 92.09 g/mol
Index Merck:13,04497
Température d'entreposage:Ambient
EINECS:200-289-5
:177
:M.O.C. Category N
Customers Who Bought This Also Bought