di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS, Macron Fine Chemicals™
# de catalogue MK238604
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 6106-24-7
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:PE bottle, Mall Polystormor®
- Size:500 g
- Cat. no.:MK238604
- Supplier No.:2386-04
Spécifications
Résultats des tests de spécification
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (Na₂C₄H₄O₆·2H₂O):99.0 - 101.0 %
- ACS - Loss on Drying at 150°C:15.61 - 15.71 %
- ACS - pH of 5% Solution at 25°C:7.0 - 9.0
- ACS - Insoluble Matter:≤0.005 %
- ACS - Chloride (Cl):≤5 ppm
- ACS - Phosphate (PO₄):≤5 ppm
- ACS - Sulfate (SO₄):≤0.005 %
- ACS - Ammonium (NH₄):≤0.003 %
- ACS - Calcium (Ca):≤0.01 %
- ACS - Heavy Metals (as Pb):≤5 ppm
- ACS - Iron (Fe):≤0.001 %
- Macron Fine Chemicals™, di-Sodium L(+)-tartrate dihydrate 99.0-101.0%, granular, AR® ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Sodium L-(+)-tartrate dihydrate,L(+)-Tartaric acid disodium salt dihydrate,(R,R)-(+)-Tartaric acid disodium salt dihydrate,(R,R)-(+)-Sodium tartrate dibasic dihydrate,Disodium L-(+)-Tartrate Dihydrate,(R,R)-(+)-Disodium tartrate dihydrate,L(+)-Tartaric acid, disodium salt, dihydrate,Sodium L-tartrate dibasic dihydrate,L(+)-Sodium tartrate dibasic dihydrate,Sodium Tartrate, Dihydrate,(+)-Disodium L-tartrate dihydrate
Point de Fusion:57 °C
Densité:1.818 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 57 °C
:Flash Pt:
:Flash Pt:
:M.W. 230.08 g/mol
Index Merck:12,08833
:di-Sodium L(+)-tartrate dihydrate
EINECS:212-773-3
:M.O.C. Category N
Customers Who Bought This Also Bought