Sulfuric acid 0.1 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # JT5641-2
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7664-93-9
MOC: Non-Regulated, With InformationCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:PE bottle, narrow mouth
- Size:1 L
- Cat. no.:JT5641-2
- Supplier No.:5641-02
Specifications
Specification Test Results
- Normality:0.0995 - 0.1005
- Chloride (Cl):= 1 ppm
- Phosphate (PO₄):= 1.0 ppm
- Heavy Metals (as Pb):= 0.3 ppm
- Trace Impurities - Iron (Fe):= 0.5 ppm
- Standardization at 25°C traceable to NIST Standard Reference Material.:
- SRM No…:
- J.T.Baker®, Sulfuric acid 0.1 N in aqueous solution, BAKER ANALYZED® volumetric solution:
Specification Test Results
About this item
Density:1,01 g/cm³ (20 °C)
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 330 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 10,38 °C
:Pt Eclair:|Point eclaire:
:M.W. 98,08 g/mol
Density:1.01 g/cm³ (20 °C)
Storage Temperature:Température ambiante
EINECS:231-639-5
:10,38
:330
UN:2796
:98.08
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 330 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 10.38 °C
:Flash Pt:
:Flash Pt:
:Density: 1.84 g/cm³ (20 °C)
:M.W. 98.08 g/mol
:GHS05
:8
:II
:1.84
:10.38
:M.O.C. Category NI
Storage Temperature:Ambient
Customers Who Bought This Also Bought