Citric acid, anhydrous 99.5-100.5% (by anhydrous basis), powder USP, J.T.Baker®
Catalog # JT0122-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 77-92-9
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Size:500 g
- Cat. no.:JT0122-1
- Supplier No.:0122-01
Specifications
Specification Test Results
- GMP Manufactured Product:
- Meets USP Requirements:
- Preserve in Tight Containers:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (HOC(COOH)(CH₂COOH)₂) (anhydrous basis):99.5 - 100.5%
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification:Passes Test
- USP - Residue on Ignition:≤0.1%
- USP - Oxalic Acid:≤0.036%
- USP - Sulfate (SO₄):≤0.015%
- USP - Readily Carbonizable Substances:Passes Test
- USP - Identification C: Water (H₂O):≤1.0%
- Must be subjected to further processing during the preparation of:
- injectable dosage forms:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Meets Reagent Specifications for testing USP/NF monographs:
- Metallic Residues: No metal catalysts or metal reagents, as defined by:
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:
- this material.:
- J.T.Baker®, Citric acid, anhydrous 99.5-100.5% (by anhydrous basis), powder USP:
Specification Test Results
About this item
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149 - 151 °C
Density:1,665 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 310 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: 149 à 151 °C
:Pt Eclair:|Point eclaire:
:M.W. 192,13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.665 g/cm³ (20 °C)
EINECS:201-069-1
:310
Storage Temperature:Température ambiante
REACH:01-2119457026-42-0000
:GHS07
:99,5-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 310 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 149…151 °C
:Flash Pt:
:Flash Pt:
:M.W. 192.13 g/mol
:M.O.C. Category R
Storage Temperature:Ambient
Customers Who Bought This Also Bought