Ethylene glycol ≥99.0%, Semi Grade for the electronics industry, VWR Chemicals BDH®
Catalog # BDH2033-1GLP
Supplier: BDH
CAS Number: 107-21-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:HDPE bottle
- Size:1 gal.
- Cat. no.:BDH2033-1GLP
- Supplier No.:10730S1G41BD
Specifications
Specification Test Results
- Assay (C6H6O2):99.0% Min.
- Color (APHA):10 APHA Max.
- Acidity:0.01% Max.
- Water (H2O):0.1% Max.
- Boiling Range:194 ºC to 200 ºC
- Specific Gravity at 25ºC:1.112 to 1.120
- Appearance:To Pass Test
- Al Impurities:100 ppb Max.
- B Impurities:100 ppb Max.
- Ca Impurities:100 ppb Max.
- Cr Impurities:100 ppb Max.
- Cu Impurities:100 ppb Max.
- Fe Impurities:100 ppb Max.
- Mg Impurities:100 ppb Max.
- Mn Impurities:100 ppb Max.
- Ni Impurities:100 ppb Max.
- K Impurities:100 ppb Max.
- Na Impurities:100 ppb Max.
- Pb Impurities:100 ppb Max.
- Sn Impurities:100 ppb Max.
- Zn Impurities:100 ppb Max.
- VWR Chemicals BDH®, Ethylene glycol ≥99.0%, Semi Grade for the electronics industry:
Specification Test Results
About this item
Boiling Pt:196…198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
Flash Pt:116 °C
:Boiling Pt:
:Boiling Pt: 196 to 198 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –13 °C
:Flash Pt:
:Flash Pt: 116 °C
:M.W. 62.07 g/mol
Merck Index:13,03832
:Ethylene glycol
:GHS07
:196…198 °C (1013 hPa)
:116
:-13
Storage Temperature:Refrigerator
EINECS:203-473-3
REACH:01-2119456816-28
Customers Who Bought This Also Bought