n-Heptane ≥99%, Laboratory Reagent, VWR Chemicals BDH®
# de catalogue BDH1127-4LG
Fournisseur: VWR International
Numéro CAS: 142-82-5
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:BDH1127-4LG
- Supplier No.:PL207ZA-4
Spécifications
Résultats des tests de spécification
- Appearance:Clear
- Water (K.F.):Less than 0.010%
- Residue:Less than 0.001% (0.0010 g/146 mL)
- GC Purity:Greater than 99.9% n-heptane (n-C7) and C7 isomers (Typical - 99.9+%)
- n-Heptane:Greater than 99.0%
- Color (APHA):Less than 10
- VWR Chemicals BDH®, n-Heptane ≥99%, Laboratory Reagent:
Résultats des tests de spécification
A propos de cet article
Point d'ébullition:98,4 °C (1013 hPa)
Point de Fusion:-90,6 °C
Densité:0,6838 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
Point d'éclair:-4 °C
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 98,4 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -90,6 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: -4 °C
:M.W. 100,20 g/mol
Index Merck:13,04679
Point d'ébullition:98.4 °C (1013 hPa)
Point de Fusion:–90.6 °C
Point d'éclair:–4 °C
Température d'entreposage:Ambient
EINECS:205-563-8
:-90,6
:98,4
Densité:0.6838 g/cm³ (20 °C)
ONU:1206
:-4
:GHS02
:GHS08
:GHS07
:GHS09
Température d'entreposage:Température ambiante
:99
:Boiling Pt:
:Boiling Pt: 98.4 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –90.6 °C
:Flash Pt:
:Flash Pt: -4 °C
:M.W. 100.20 g/mol
:-90.6
:3
:II
Customers Who Bought This Also Bought