2-Mercaptoethanol ≥99% for biotechnology
Catalog # 97064-588
Supplier: VWR
CAS Number: 60-24-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Bottle, amber
- Size:100 ml
- Cat. no.:97064-588
- Supplier No.:0482-100ML
Specifications
Specification Test Results
- DNase:none detected
- Protease:none
- Purity:99 %
- RNase:none detected
- Water:= 0.5 %
- 2-Mercaptoethanol ≥99% for biotechnology:
Specification Test Results
About this item
:Mercapto-2 éthanol
Boiling Pt:157,2 °C
Melting Pt:-100 deg °C
Density:1,12 g/cm³ (20 °C)
ADR:6.1,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
Flash Pt:68 °C
:Pt ébul.:|Point d'ébullition:
:Pt ébul.: 157 °C (1013 hPa)
:Pt Fusion:|Point de fusion:
:Pt Fusion: -100 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: 68 °C
:M.W. 78,14 g/mol
Merck Index:13,05894
Boiling Pt:157.2 °C
EINECS:200-464-6
:-100
:157
Melting Pt:–100 deg °C
UN:2966
:68
:GHS06
:GHS08
:GHS05
:GHS07
Density:1.12 g/cm³ (20 °C)
Flash Pt:68 °C
Storage Temperature:Température ambiante
:99
:Boiling Pt:
:Boiling Pt: 157 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –100 deg °C
:Flash Pt:
:Flash Pt: 68 °C
:M.W. 78.14 g/mol
:2-Mercaptoethanol
:6.1
:II
Storage Temperature:Ambient
Customers Who Bought This Also Bought