Trypan blue 0.4% in aqueous solution ready-to-use, sterile
# de catalogue 97063-702
Fournisseur: VWR
Product available on GSA Advantage®
Numéro CAS: 72-57-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Size:100 mL
- Cat. no.:97063-702
- Supplier No.:K940-100ML
Spécifications
Résultats des tests de spécification
- Abs.@ Lambda Max (1:100, Water):1 - 1.3
- Lambda Max (1:100, Water):585 nm - 595 nm
- Trypan blue 0.4% in aqueous solution ready-to-use, sterile:
Résultats des tests de spécification
A propos de cet article
Synonymes: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt,CI 23850,3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt,Benzamine blue,Trypan blue sodium salt,Direct Blue 14,AzidinBlue,Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Eclair:|Point eclaire:
:M.W. 960,82 g/mol
:GHS08
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 960.82 g/mol
Customers Who Bought This Also Bought