SPILFYTER® KOLORSAFE® Acid and Base Neutralizers, NPS
# de catalogue 56616-404
Fournisseur: NPS
Spécifications
- Description:Acid Neutralizer
- Size:0.9 L (0.25 gal.)
- Supplier No.:410001
Spécifications
A propos de cet article
Neutralize hazardous caustic and acid spills (including hydrofluoric acid) with these easy-to-use liquid neutralizers.
- Kits are available in dry or liquid formulas
- Sturdy yellow bucket keeps contents clean and dry when not in use
Color change indicates when enough neutralizer has been applied to the spill. Particulate-free for cleanroom applications. Available in 0.9 L (0.25 gal.), 3.8 L (1 gal.), and 18.9 L (5 gal.) sizes.
Delivery: Kit includes 7.6 cm x 122 cm Hazmat Mini-Booms (3), 30 cm x 30 cm Hazmat Pads (10), 3.8 L KOLOR-SAFE® Dry or Liquid Acid/Base Neutralizer (1), Chemical Classifier Strips (2), Chemical Classifier Chart (1), Nitrile Gloves (1 pr.), Splash Goggles (1 pr.) and Disposal Bag (1).
Packaging: Spill Kits are packed in a 5-gallon plastic bucket for easy portability and disposal of spill clean-up materials.
Customers Who Bought This Also Bought