ContactUsLinkComponent
Fendall® Saline Eyewash Wall Stations, Honeywell Safety
  56611-128
undefined
Fendall® Saline Eyewash Wall Stations, Honeywell Safety
  56611-128
 :  32-000461-0000
  • Description:
    Single station
  • Size:
    950 ml (32 oz.)
  • Cat. no.:
    56611-128
  • Supplier No.:
    32-000461-0000
Available in 470 ml (16 oz.) and 950 ml (32 oz.) single and 950 ml (32 oz.) double wall stations.
  • Two-sided tape and screws are provided for mounting
  • Classified as personal/secondary eyewash device
Bottles have extended flow openings and twist-off tops for easy access in an emergency. Each bottle is expiry dated and lot numbered for end-user protection.
Frequently Bought Together