Order Entry
ContactUsLinkComponent
BUFFER 220MV REDOX 250ML
Redox Buffer Solution, Mettler-Toledo
# de catalogue 11238-454
Fournisseur:  Mettler Toledo
Product available on GSA Advantage®
Numéro CAS:  
BUFFER 220MV REDOX 250ML
Redox Buffer Solution, Mettler-Toledo
# de catalogue 11238-454
Fournisseur:  Mettler Toledo
Product available on GSA Advantage®
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Redox Buffer Solution, 220 mV/pH 7
  • Size:
    250 mL
  • Reference Temperature:
    25 °C
  • Cat. no.:
    11238-454
  • Supplier No.:
    51350060

Spécifications

A propos de cet article

Redox buffer solutions are used for verification of all common redox sensors

  • Temperature dependent
  • Used in quality assurance systems
Delivery: Supplied with a quality inspection certificate.
Customers Who Bought This Also Bought