Spécifications
- Description:Epoxy Refillable Triode
- Cat. no.:10010-738
- Supplier No.:9180BNMD
Spécifications
A propos de cet article
Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance
- Available in epoxy triode low maintenance and epoxy refillable triode construction
- Built-in thermistor for Automatic Temperature Compensation (ATC)
- Not designed for amperometric or constant current titrations, such as Karl Fischer analysis
Orion™ Redox/ORP/Temp Electrodes measure ORP and temperature using a platinum redox sensor, a silver/silver chloride internal reference system, and a built-in thermistor which is adapted for Automatic Temperature Compensation (ATC).