Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
# de catalogue JT0115-1
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Numéro CAS: 5949-29-1
MOC : RéglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle, amber, wide mouth, black T/E cap
- Size:500 g
- Supplier No.:0115-01
Spécifications
Résultats des tests de spécification
- GMP Manufactured Product:
- Meets BP Chemical Specifications:
- Meets EP Chemical Specifications:
- Meets JP Chemical Specifications:
- Meets USP Requirements:
- Preserve in Tight Containers:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification:Passes Test
- USP - Water (H₂O):7.5 - 9.0%
- USP - Readily Carbonizable Substances:Passes Test
- USP - Residue on Ignition:≤0.1%
- USP - Sulfate (SO₄):≤0.015%
- USP - Oxalic Acid:≤0.036%
- USP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5%
- EP/BP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5%
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Readily Carbonizable Substances:Passes Test
- EP/BP - Oxalic Acid:≤ 360 ppm
- EP/BP - Sulfate (SO₄):≤ 150 ppm
- EP/BP - Water (H₂O):7.5 - 9.0%
- EP/BP - Ash (sulfated):≤0.1%
- EP/BP - Endotoxin Concentration, IU/mg:≤0.5
- JP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5%
- JP - Identification:Passes Test
- JP - Sulfate (SO₄):≤150 ppm
- JP - Oxalic Acid:≤360 ppm
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Water (H₂O):7.5 - 9.0%
- JP - Readily Carbonizable Substances:Passes Test
- JP - Residue on Ignition:≤0.10%
- Must be subjected to further processing during the preparation of:
- injectable dosage forms.:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial:
Résultats des tests de spécification
A propos de cet article
Synonymes: Hydroxytricarballylic acid,2-Hydroxy-1,2
:Acide citrique monohydraté
Point de Fusion:135 - 152 °C
Densité:1,552 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
Point d'éclair:173,9 °C (closed cup)
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Fusion: 135 à 152 °C
:Pt Eclair:|Point eclaire:
:Pt Eclair: 173,9 °C (coupe fermée)
:M.W. 210,14 g/mol
Index Merck:13,02350
Densité:1.552 g/cm³ (20 °C)
Température d'entreposage:Ambient
EINECS:201-069-1
:173,9
:GHS07
Point de Fusion:135…152 °C
Point d'éclair:173.9 °C (closed cup)
Température d'entreposage:Température ambiante
:99,5-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 135…152 °C
:Flash Pt:
:Flash Pt: 173.9 °C (closed cup)
:M.W. 210.14 g/mol
:Citric acid monohydrate
:173.9
:M.O.C. Category R
:12,04467
:14,04459
Customers Who Bought This Also Bought