Tetrahydrofuran ≥99% stabilized ACS, VWR Chemicals BDH®
# de catalogue BDH1149-4LG
Fournisseur: VWR International
Numéro CAS: 109-99-9
MOC : Non réglementéCertificats
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Packaging:Glass bottle
- Size:4 L
- Supplier No.:PL341ZA-4
Spécifications
Résultats des tests de spécification
- Appearance:Clear, colorless liquid
- Water (K.F.):Less than 0.050%
- Residue:Less than 0.030% (0.0060/ 22.5mL)
- GC Purity:Greater than 99.0%
- Peroxides:Less than 0.015% as H2O2 (No more than 3.9 mL of 0.1N sodium thiosulfate solution)
- Color (APHA):Less than 20
- Preservative (BHT) one cm cell wavelength, nm 278:Absorbance between 1.95 and 3.12 (250 - 400 ppm)
- BHT Preservative:250 – 400 ppm
- VWR Chemicals BDH®, Tetrahydrofuran ≥99% stabilized ACS:
Résultats des tests de spécification
A propos de cet article
Synonymes: Butylene oxide,THF
Point d'ébullition:66 °C (1013 hPa)
Point de Fusion:–108.5 °C
Densité:0.888 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
Point d'éclair:–21.5 °C
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:Boiling Pt:
:Boiling Pt: 66 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –108.5 °C
:Flash Pt:
:Flash Pt: -21.5 °C
:M.W. 72.11 g/mol
Index Merck:12,09351
EINECS:203-726-8
:-108.5
Température d'entreposage:Ambient
ONU:2056
:05-2114615188-48
:3
:II
:-21.5
:GHS02
:GHS08
:GHS07
:BHT
Customers Who Bought This Also Bought