Spécifications
- Description:EZ-Vision® One
- Environmentally Preferable:Safer Human and Environmental Health
- Size:5 x 1 mL
- Supplier No.:N472-KIT
Spécifications
A propos de cet article
Provides a safe, non-mutagenic alternative to ethidium bromide for instantaneous fluorescent DNA band visualization.
- Easy and Convenient
- Instant Results
- Eliminates Hazardous Ethidium Bromide Use
This environmentally friendly dye requires no special handling or disposal, and will not incur hazardous shipping, storage, or waste charges. Supplied in a 6X loading buffer that is added to the DNA sample and can be run on a gel and visualized immediately on a UV transilluminator. Requires no post electrophoresis staining or destaining and has a very broad emission spectra with a peak near 450nm that works with most existing filters for gel documentation.
Three different tracking dye options are available. Each version contains the same non-hazardous fluorescent DNA dye and loading buffer. EZ-Vision® One is one tracking dye migrating at approximately 10bp in a 1% agarose gel. EZ-Vision® Two is two tracking dyes migrating at approximately 4,000bp and 400bp in a 1% agarose gel. EZ-Vision® Three is three tracking dyes migrating at approximately 4,000bp, 400bp, and 10bp in a 1% agarose gel.