ContactUsLinkComponent
EZ-VISION ONE DNA DYE 6X KIT 5X1ML
VWR® EZ-Vision® Dye-as-Loading Buffer, 6X
# de catalogue 97064-190
Fournisseur:  VWR
   Safer Human and Environmental Health
Numéro CAS:  
EZ-VISION ONE DNA DYE 6X KIT 5X1ML
VWR® EZ-Vision® Dye-as-Loading Buffer, 6X
# de catalogue 97064-190
Fournisseur:  VWR
   Safer Human and Environmental Health
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    EZ-Vision® One
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Size:
    5 x 1 mL
  • Supplier No.:
    N472-KIT
Spécifications
A propos de cet article
Provides a safe, non-mutagenic alternative to ethidium bromide for instantaneous fluorescent DNA band visualization.
  • Easy and Convenient
  • Instant Results
  • Eliminates Hazardous Ethidium Bromide Use
This environmentally friendly dye requires no special handling or disposal, and will not incur hazardous shipping, storage, or waste charges. Supplied in a 6X loading buffer that is added to the DNA sample and can be run on a gel and visualized immediately on a UV transilluminator. Requires no post electrophoresis staining or destaining and has a very broad emission spectra with a peak near 450nm that works with most existing filters for gel documentation.
Three different tracking dye options are available. Each version contains the same non-hazardous fluorescent DNA dye and loading buffer. EZ-Vision® One is one tracking dye migrating at approximately 10bp in a 1% agarose gel. EZ-Vision® Two is two tracking dyes migrating at approximately 4,000bp and 400bp in a 1% agarose gel. EZ-Vision® Three is three tracking dyes migrating at approximately 4,000bp, 400bp, and 10bp in a 1% agarose gel.
Customers Who Bought This Also Bought