Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Catalog # 97063-136
Supplier: VWR
Product available on GSA Advantage®
CAS Number: 67-68-5
Specifications
- Size:5×10 ml
- Supplier No.:N182-5X10ML
Specifications
Specification Test Results
- Acidity Test:Pass
- Congealing Point:= 18.3°C
- Appearance:Clear colorless liquid.
- Nonvolatile Residue:= 5 mg
- Product Lock Test:C9481
- Purity:= 99.9 %
- Refractive Index:1.4755 - 1.4775
- Specific Gravity:1.095 - 1.101
- Sterility:Pass
- Water (KF):= 0.1 %
Specification Test Results
About this item
Synonyms: Methanesulphinylmethane,Methyl sulphoxide,Dimethyl(oxido)sulphur
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
Merck Index:13,03285
Storage Temperature:Ambient
EINECS:200-664-3
Frequently Bought Together