Order Entry
ContactUsLinkComponent
BUFFER TRIS-BORATE-EDTA UL-PR GRD 4L
VWR® TRIS-Borate-EDTA Buffer, 10X Liquid Concentrate, Ultra Pure Grade
# de catalogue 97062-122
Fournisseur:  VWR
Numéro CAS:  
BUFFER TRIS-BORATE-EDTA UL-PR GRD 4L
VWR® TRIS-Borate-EDTA Buffer, 10X Liquid Concentrate, Ultra Pure Grade
# de catalogue 97062-122
Fournisseur:  VWR
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Size:
    4 L
  • Cat. no.:
    97062-122
  • Supplier No.:
    0658-4L

Spécifications

A propos de cet article

TBE is optimized for DNA electrophoresis techniques using both acrylamide and agarose.A concentrated (10X), pH adjusted solution of Ultra Pure Grade Tris Buffer, ACS Grade Boric Acid & EDTA which when reconstituted to a working (1X) Solution concentration, contains 0.089M Tris, 0.089M Borate & 0.002M EDTA.

  • High Buffering Capacity
  • High Ionic Strength
  • Permits Extended Electrophoresis Runs with no Buffer Recirculation
  • Convenient Packaging Options Minimize Weighing & Mixing

*Note: TBE has a tendency to precipitate over time. If this occurs, redissolve in solution.

No DNase, Protease, RNase. pH (1:10) @25°C is 8.2 - 8.4. Conductivity (1:10) @25°C is 750-1250 umhos.

Customers Who Bought This Also Bought