To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Ø:16 mm
- Volume:9 ml
- Description:K3EDTA Tube
- Length:100 mm
- Cat. no.:95057-237
- Tube Dimensions:16 x 100 mm
- Ring Color:Black
- Cap Color:Lavender
- Supplier No.:455036
Specifications
About this item
Designed for use in the examination of whole blood in hematology applications, EDTA tubes feature interior walls that are coated with either K2EDTA or K3EDTA.
- Gel separator forms stable barrier between serum and blood cells
- PET plastic construction is virtually unbreakable
- Color-coded rings in caps
The EDTA binds calcium ions and blocks the coagualtion cascade
The EDTA tubes equipped with a gel separator contain a barrier gel in the base of the tube. During centrifugation, this gel forms a stable barrier between the serum and the blood cells, separating the serum from fibrin and cells.
VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with non-ridged pull caps that feature an exterior color-coded ring to indicate either the additive used or draw volume. A black ring indicates standard tubes, a yellow ring indicates the inclusion of a separation gel, and a white ring indicates a tube designed to accommodate pediatric draw.
Tubes are spray-dried with 1.2–2.0mg EDTA/1 mL of blood. Tubes can also be used in the closed mode with direct sampling analyzers.