Order Entry
ContactUsLinkComponent
Blood Collection Tube 2.7 ml 3.2.% Na Cit. pk50
VACUETTE® Sodium Citrate Coagulation Tubes, Greiner Bio-One
Catalog # 95057-171
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
Blood Collection Tube 2.7 ml 3.2.% Na Cit. pk50
VACUETTE® Sodium Citrate Coagulation Tubes, Greiner Bio-One
Catalog # 95057-171
Supplier:  Greiner Bio-One
Product available on GSA Advantage®
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Ø:
    13 mm
  • Volume:
    3 ml
  • Description:
    Tube with 3.2% (0.109M) Sodium Citrate Solution
  • Length:
    75 mm
  • Cat. no.:
    95057-171
  • Tube Dimensions:
    13 x 75 mm
  • Ring Color:
    Black
  • Supplier No.:
    454334

Specifications

About this item

Designed for use in the examination of coagulation parameters, coagulation tubes contain a buffered tri-sodium citrate solution.

  • PET plastic construction is virtually unbreakable
  • Contains buffered tri-sodium citrate solution
  • Color-coded rings in caps indicate draw volume

Citrate concentrations of either 3.2% (0.109M) or 3.8% (0.129M) are available, with a mixing ratio of one part citrate to nine parts blood.

VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with blue non-ridged pull caps that feature an exterior color-coded ring to indicate the draw volume. A black ring indicates standard tubes and a white ring indicates a tube designed to accommodate pediatric draw. High altitude tubes are recommended for use in locations 5,000ft. or higher above sea level.

Customers Who Bought This Also Bought