To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- ā¢ State issued document with your organization's Federal Tax ID Number
- ā¢ State issued document with your organization's Resale Tax ID Number
- ā¢ City or County issued Business License
- ā¢ State Department of Health Services License
- ā¢ Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
SpƩcifications
- Description:Feather Double Edge Blade
- Supplier No.:72002-01
SpƩcifications
A propos de cet article
Made from carbon steel.
FeatherĀ® blades are well known to have super sharp edges. Easily breaks in half for use with any tissue slicing machine, or break smaller portions for use as a microscalpel with a Blade Breaker and Holder. Blades are available in packages of 10 blades per box or 500 blades per box. Measurements W x L x T: 21.95 x 42.96 x 0.10 mm (0.865 x 1.69 x 0.004")