ContactUsLinkComponent
ACRYLIC RBM ADULT MOUSE, CORONAL
The Economy Rodent Brain Matrix, Electron Microscopy Sciences
# de catalogue 100492-848
Minority or Woman-Owned Business Enterprise
ACRYLIC RBM ADULT MOUSE, CORONAL
The Economy Rodent Brain Matrix, Electron Microscopy Sciences
# de catalogue 100492-848
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Acrylic Rodent Brain Matrix Adult Mouse, Coronal
  • Size:
  • Supplier No.:
    69080-C
Spécifications
A propos de cet article
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
  • Highly polished
  • Precision machined
  • Precise blocking of the brain
  • Reproducible 1 mm segments
  • Cost saving and in all directions
Customers Who Bought This Also Bought