Spécifications
- Description:Acrylic Rodent Brain Matrix Adult Mouse, Coronal
- Size:—
- Supplier No.:69080-C
Spécifications
A propos de cet article
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
- Highly polished
- Precision machined
- Precise blocking of the brain
- Reproducible 1 mm segments
- Cost saving and in all directions