Spécifications
- Description:Paper Points, Selection 1
- Cat. no.:100500-760
- Supplier No.:71011-01
Spécifications
A propos de cet article
Machine rolled paper with a tapered end which offers good water absorbency, and assists you in the removal of surplus water from the knife boat, on the block face, and from the grids. Available in two assortment sizes. The bigger the number the larger the point.