Specifications
- Description:Plastic Canoe Medium
- Cat. no.:100488-958
- Supplier No.:70043-20
Specifications
About this item
Flexible dishes for handling liquids or solids.
Ideal for controlled pour-off. These Anti-Static Canoes with contoured sides provide a convenient non-slip gripping surface. As well, they allow for more accurate weighing and reading of a sample. The canoe shape enables pouring of weighed samples.