Plastic Forceps, with Jaw Grips, Electron Microscopy Sciences
Product Details & Documents
These plastic forceps are excellent for handling hot, cold, corrosive or contaminated objects. Made from blue polypropylene with interlocking teeth.
A catch locks tips tightly for use as a clamp. Scissor type handles with double action hinges makes this forcep a useful cryo tool, and great for handling stubs.
Spécifications
- Color:Blue
- Désignation:Plastic Forcep with Jaw-Grips