ContactUsLinkComponent
Polyethylene wafer tweezers
Plastic Tweezers and Tweezers Accessories, Electron Microscopy Sciences
Catalog # 100494-696
Minority or Woman-Owned Business Enterprise
Polyethylene wafer tweezers
Plastic Tweezers and Tweezers Accessories, Electron Microscopy Sciences
Catalog # 100494-696
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Polyethylene Wafer, Extra Wide
  • Material:
    Linear polyethylene
  • Tip Configuration:
    With extra wide point
  • Cat. no.:
    100494-696
  • Overall Length:
    11.3 cm
  • Supplier No.:
    78310-71
Specifications
About this item
Plastic wafer tweezers are low cost and designed for ease of use. Available in glass filled Delrin and acid resistant fluorocarbon or polyethylene.
Customers Who Bought This Also Bought