Spécifications
- Sterility:Non Sterile
- Volume:10 ml
- Description:Sample Vials with White Caps
- Supplier No.:63034-10
Spécifications
A propos de cet article
Transport vials/tubes are ideal for sample collection and storage.
- Conical tubes with freestanding skirts
- Tubes are autoclavable
- Write-on area on side
Transport Vials are made from virgin polypropylene. They are available in three sizes, and come either sterile or non sterile. Graduations provide convenient and accurate references.
Transport Tubes are made from virgin polypropylene (PP) or polystyrene (PS). They also work well in Olympus automated testing equipment. Plug caps are required.