To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Numbering:#1
- Volume:1 oz.
- Description:Cover glasses, Super Slip, rectangular
- L×W:60×24 mm
- Packaging:80 pieces/oz.
- Supplier No.:72236-60
Spécifications
A propos de cet article
Finally, your automated coverslipper has the right coverslip to allow it to run smoothly. These “Superslip®” coverslips are designed to work in automatic coverslip machines and humid environments without sticking or jamming up in the middle of a run.
These new superslips combine specialized production procedures with our unique moisture-resistant packaging which brings you superior coverslips.