Specifications
- Description:Instant Readout Digital Caliper
- Cat. no.:100502-462
- Supplier No.:62065-40
Specifications
About this item
This caliper is made from stainless steel and measures inside and outside in inches and millimeters.
Read-out from 0 to 6 inches with 0.001" resolution, or 0 to 150 mm with 0.01 mm resolution. Includes a zero set button, depth measuring rod and a fine adjustment thumb wheel.