Spécifications
- Description:S-50 Sedgewick Rafter, Plastic
- Supplier No.:68050-50
Spécifications
A propos de cet article
For use in water analysis, culture inspection, and any fluid where particles per unit volume need to be determined. Available in two versions.
S50 is an economically priced plastic cell. S52 is made from glass, with a chromium surface image. This is intended for professional use and when using phase contrast. In each type, a central cell is filled with fluid and a coverglass limits the volume to 1 milliliter. The grid subdivides this into micro-liters. S51 is the spare cover glass for S50 and S52. Each cell is supplied with one cover glass.