ContactUsLinkComponent
S-50 SEDGEWICK RAFTER, PLASTIC
Sedgewick Rafter (Microlite) Cell, Electron Microscopy Sciences
# de catalogue 100499-392
Minority or Woman-Owned Business Enterprise
S-50 SEDGEWICK RAFTER, PLASTIC
Sedgewick Rafter (Microlite) Cell, Electron Microscopy Sciences
# de catalogue 100499-392
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    S-50 Sedgewick Rafter, Plastic
  • Supplier No.:
    68050-50
Spécifications
A propos de cet article
For use in water analysis, culture inspection, and any fluid where particles per unit volume need to be determined. Available in two versions.
S50 is an economically priced plastic cell. S52 is made from glass, with a chromium surface image. This is intended for professional use and when using phase contrast. In each type, a central cell is filled with fluid and a coverglass limits the volume to 1 milliliter. The grid subdivides this into micro-liters. S51 is the spare cover glass for S50 and S52. Each cell is supplied with one cover glass.
Customers Who Bought This Also Bought