ContactUsLinkComponent
Low-temp hot glue gun
Low Temperature Hot Glue Gun, Electron Microscopy Sciences
# de catalogue 100493-644
Minority or Woman-Owned Business Enterprise
Numéro CAS:  
Low-temp hot glue gun
Low Temperature Hot Glue Gun, Electron Microscopy Sciences
# de catalogue 100493-644
Minority or Woman-Owned Business Enterprise
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Low-Temp Hot Glue Gun
  • Supplier No.:
    72171-00
Spécifications
A propos de cet article
This low temperature mini glue gun is safe and allows for the easy application of the melt glue to the aluminum stub for the mounting of bulk or irregular shaped specimens for SEM.
  • Low temperature
  • Easy to use
  • 40" cord
Comes complete with a 40" cord and two glue sticks. Accepts 5/16" diameter all temperature or low temperature glue sticks. 110 volt, 10 watts.
Customers Who Bought This Also Bought