Spécifications
- Description:Low-Temp Hot Glue Gun
- Supplier No.:72171-00
Spécifications
A propos de cet article
This low temperature mini glue gun is safe and allows for the easy application of the melt glue to the aluminum stub for the mounting of bulk or irregular shaped specimens for SEM.
- Low temperature
- Easy to use
- 40" cord
Comes complete with a 40" cord and two glue sticks. Accepts 5/16" diameter all temperature or low temperature glue sticks. 110 volt, 10 watts.