To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Dimensions:25 x 32 mm
- Color:Blue
- Description:Biopsy Foam Pad
- For Use With:1 x 1 1/4" Casettes
- Supplier No.:62325-06
Spécifications
A propos de cet article
A cellular polyester blue foam that has uniform air pockets throughout the pad for optimal fluid flow
It prevents either small or fragmented tissue specimens from being lost. To Use: Place small samples or specimen fragments between the two pads and place the specimen in a cassette or capsule.