Order Entry
ContactUsLinkComponent
VWR* Nylon Test Tube Rack 20Mm 40-Places - Blue
VWR® Test Tube Racks
Catalog # 89215-778
Supplier:  VWR International
VWR® Test Tube Racks
Catalog # 89215-778
Supplier:  VWR International

Specifications

  • No. of Holes:
    40
  • Color:
    Blue
  • Dimensions:
    25.4L X 10.4W X 8.7H cm (10 x 43/32 x 37/16")
  • Cat. no.:
    89215-778
  • For Tube O.D.:
    20 mm

Specifications

About this item

VWR® Test Tube Racks are available in both nylon glass filled or polypropylene options.

  • Both glass filled or polypropylene options are easy to handle
  • Racks may be used in an incubator, fume hood, refrigerator, or for benchtop applications
  • Autoclavable
  • No assembly required

Racks are a single piece molded from either nylon 6 glass fiber reinforced material, or polypropylene.

Customers Who Bought This Also Bought