Order Entry
ContactUsLinkComponent
VWR* Nylon Test Tube Rack 16Mm 72-Places - Yellow
VWR® Test Tube Racks
Catalog # 89215-776
Supplier:  VWR International
VWR* Nylon Test Tube Rack 16Mm 72-Places - Yellow
VWR® Test Tube Racks
Catalog # 89215-776
Supplier:  VWR International

Specifications

  • No. of Holes:
    72
  • Color:
    Yellow
  • Dimensions:
    25.1L X 12.7W X 7.3H cm (97/8 x 5 x 27/8")
  • Cat. no.:
    89215-776
  • For Tube O.D.:
    16 mm

Specifications

About this item

VWR® Test Tube Racks are available in both nylon glass filled or polypropylene options.

  • Both glass filled or polypropylene options are easy to handle
  • Racks may be used in an incubator, fume hood, refrigerator, or for benchtop applications
  • Autoclavable
  • No assembly required

Racks are a single piece molded from either nylon 6 glass fiber reinforced material, or polypropylene.

Customers Who Bought This Also Bought