ContactUsLinkComponent
Reactor Body Jacketed 500Ml
Photochemical Jacketed Reactor with Ace-Thred (No Valve), Ace Glass Incorporated
# de catalogue 89215-148
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Reactor Body Jacketed 500Ml
Photochemical Jacketed Reactor with Ace-Thred (No Valve), Ace Glass Incorporated
# de catalogue 89215-148
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Capacity:
    500 ml
  • Cat. no.:
    89215-148
  • Supplier No.:
    7864-10
Spécifications
A propos de cet article
Jacketed reaction vessel designed to enable cooling of reactant material during photolysis
Constructed of borosilicate glass to accommodate the immersion wells. With one standard taper 14/20 angled joint for sparger tube, one standard taper 24/40 vertical joint for condenser and one #7-Ace-Thred side arm for thermometer. Center joint is #50 Ace-Thred. Bottom of reactor is flat to allow use of stir bar. Volumes indicated are total volumes. Volume in reactive area of lamp is approximately 40-50% of total volume. Size C hose connections. Listing is vessel only.
Customers Who Bought This Also Bought