ContactUsLinkComponent
Cap Insert 12.0Mm Id
Duran Bottle GL 80 Connection System, Ace Glass Incorporated
# de catalogue 89214-934
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Cap Insert 12.0Mm Id
Duran Bottle GL 80 Connection System, Ace Glass Incorporated
# de catalogue 89214-934
Fournisseur:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
  • Description:
    Insert for GL18 Screw Cap, 12 mm I.D.
  • Cat. no.:
    89214-934
  • Supplier No.:
    7631-14
Spécifications
A propos de cet article
The heart of the GL 80 connection system is a GL80 polypropylene cap with 4, GL18 threaded ports
Modular accessories include polypropylene tube connection caps,solid polybutylene terepthalate (PBT) caps, PBT pressure equalization caps and PTFE cap inserts to accommodate various sizes of tubing. A typical application is the safe transfer of liquid media within a closed and sterile system for which you'd need one two-pack code 89214-928 GL80 cap, two 89214-906 equalization caps, and some combination of solid caps and tubing connections with inserts
Customers Who Bought This Also Bought